Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 250 x 4.6mm

Catalog No. 502394372 Shop All Phenomenex Products
Encompass

Unisol 3m C18 100 UO932505-0

Catalog No. 50-239-4372 Supplier Phenomenex Inc Supplier No. UO9325050
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Unisol 3m C18 100 UO932505-0

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.