Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Sorbent, 100g/Vial

Catalog No. 502394630 Shop All Phenomenex Products
Encompass

Unisol 5m Amide 100 VH950100-0

Catalog No. 50-239-4630 Supplier Phenomenex Inc Supplier No. VH9501000
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Unisol 5m Amide 100 VH950100-0

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.