Learn More
Imtakt UNISON UK-AMINO HPLC COLUMN
Catalog No. NC2226479
Encompass
Inner Diameter: 2.0mm, Length: 150mm, Pressure: Standard Pressure
This item is not returnable. View return policy
Catalog No. NC2226479 Supplier Imtakt Supplier No. UKA25
Inner Diameter: 2.0mm, Length: 150mm, Pressure: Standard Pressure