Learn More
Imtakt UNISON UK-C18 3UM 4.6X250
Catalog No. NC9986004
Encompass
UNISON UK-C18. 100% aqueous stable 3� C18 column. 4.6mm x 250. The Unison Family of columns provide a wide range of high quality reverse phase and normal phase columns. The quality of our material provides high efficiency with low backpressure and 100% aqueous stability. Peak shape is also improved with the use of our polymeric end-capping technology. The Unison series of columns are available with C18, C8, Phenyl, Amino and Silica stationary phases to meet a wide range of chromatographic needs.
Catalog No. NC9986004 Supplier Imtakt Supplier No. UK006
UNISON UK-C18. 100% aqueous stable 3� C18 column. 4.6mm x 250. The Unison Family of columns provide a wide range of high quality reverse phase and normal phase columns. The quality of our material provides high efficiency with low backpressure and 100% aqueous stability. Peak shape is also improved with the use of our polymeric end-capping technology. The Unison series of columns are available with C18, C8, Phenyl, Amino and Silica stationary phases to meet a wide range of chromatographic needs.