Learn More
Waters Corp UniSpray UPC2 Probe Assembly, 750 mm x 50 μm, ESI
Catalog No. 502391846 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement UniSpray UPC2 Probe Assembly. It features a tool-free design and is used on universal source designs. This ESI Probe Assembly measures 750 mm x 50 μm.
  • Tool-free ESI Probe Assembly
  • Used on universal source designs
Catalog No. 50-239-1846 Supplier Waters Corp Supplier No. 700011373
This is a replacement UniSpray UPC2 Probe Assembly. It features a tool-free design and is used on universal source designs. This ESI Probe Assembly measures 750 mm x 50 μm.
  • Tool-free ESI Probe Assembly
  • Used on universal source designs