Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Owen Mumford™ Unistik™ Pro Single Use Safety Lancet
SDP

Catalog No. 22024103 Shop All Owen Mumford, Inc. Products
Change view
Click to view available options
Flow Type:
High
Medium
Needle Gauge:
21 G
25 G
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Type Needle Gauge
22-024-103 Medium 25 G
22-024-102 High 21 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-024-103 Supplier Owen Mumford™ Supplier No. AT 1634

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Featuring a syringe-like grip and a push-button design, Unistik™ Pro offers a reliable blood sampling solution that’s ergonomic, intuitive and affordable.

  • Comfort Zone™ Technology for enhanced patient comfort
  • Intuitive design makes blood sampling easy for healthcare professionals
  • Needle is hidden before use and permanently retracts after use to help protect against accidental needle sticks and cross contamination

Specifications

Disposable Single-use
Sterility Sterile
For Use With (Application) Capillary Blood Sampling
Depth (Metric) 1.6 mm
Needle Gauge 25 G
Flow Type Medium
Style Top Activated
Product Type Unistik™ Pro Safety Lancet
Color Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.