Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Owen Mumford™ Unistik™ Pro Single Use Safety Lancet
SDP

Catalog No. p-7201203 Shop All Owen Mumford, Inc. Products
Change view
Click to view available options
Flow Type:
High
Medium
Needle Gauge:
21 G
25 G
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Type Needle Gauge
22-024-102 High 21 G
22-024-103 Medium 25 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-024-102 Supplier Owen Mumford™ Supplier No. AT 1614

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Featuring a syringe-like grip and a push-button design, Unistik™ Pro offers a reliable blood sampling solution that’s ergonomic, intuitive and affordable.

  • Comfort Zone™ Technology for enhanced patient comfort
  • Intuitive design makes blood sampling easy for healthcare professionals
  • Needle is hidden before use and permanently retracts after use to help protect against accidental needle sticks and cross contamination

Specifications

Needle Gauge 21 G
Disposable Single-use
Depth (Metric) 2.0 mm
Product Type Unistik™ Pro Safety Lancet
Flow Type High
Style Top Activated
For Use With (Application) Capillary Blood Sampling
Sterility Sterile
Color Pink
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.