Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Owen Mumford™ Unistik™ Touch Single Use Safety Lancet
SDP

Catalog No. 22024105 Shop All Owen Mumford, Inc. Products
Change view
Click to view available options
Flow Type:
High
Low
Lowest
Medium
Needle Gauge:
21 G
23 G
28 G
30 G
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Type Needle Gauge
22-024-105 High 21 G
22-024-106 Medium 23 G
22-024-107 Low 28 G
22-024-108 Lowest 30 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-024-105 Supplier Owen Mumford™ Supplier No. AT 1414
Add to Cart
Add to Cart
This item is not returnable. View return policy

Designed to make blood sampling easy for the healthcare professional and more comfortable for the patient. Touch activation ensures that the device requires minimal training and can be used in three easy steps.

  • Greater comfort- Features Comfort Zone Technology™ designed to reduce pain and enhance patient comfort during sampling
  • Peace of Mind-Individually marked lot numbers for complete traceability
  • Broad range of sizes to meet different testing requirements
  • Permanently retracts to help protect against accidental needle sticks and cross contamination

Specifications

Disposable Single-use
Needle Gauge 21 G
Style Contact Activated
Sterility Sterile
Flow Type High
Packaging Quantity 200/Pk.
Product Type Unistik™ Touch Safety Lancet
Color Orange
For Use With (Application) Capillary Blood Sampling
Depth (Metric) 2.0 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.