Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZOLL™ Unit Automatic Powerheart™ Defibrillation Pads Contact

Catalog No. 17478084 Shop All Zoll Medical Corporation Products
Change view
Click to view available options
Product Type:
Pads, Defibrillation
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-478-084 Pads, Defibrillation
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-478-084 Supplier ZOLL™ Supplier No. G5A80CS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Powerheart™ G5 fully automated with CPR English and Spanish battery pads

  • Dimensions: 3.4 x 9 x 11.8 in.
  • Operation type: Automatic
  • Power source: Battery

Specifications

Product Type Pads, Defibrillation
For Use With (Equipment) AED Unit
Weight (English) 5.7 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.