Learn More
3M™ Unitech™ Rugged Handheld Computer (Android)
Catalog No. 19071129 Shop All 3M Products
Change view
Click to view available options
Product Type:
Handheld Computer
Catalog No. Product Type
19-071-129 Handheld Computer
1 options
Catalog No. 19-071-129 Supplier 3M™ Supplier No. 7010301972
3M™ Unitech™ PA720 Rugged Handheld Computer (Android) is a powerful and versatile data collection tool with a rugged design
  • Approved Unitek™ Integrated scanner and HF/NFC/UHF RFID Reader
  • Advanced performance with industrial grade 1.5 Ghz Dual Core Processor
  • Large 4.7-inch color TFT multi-touch display
  • Gorilla™ Glass screen with 7H hardness
  • Rugged Construction: IP65 rated and 4 drop certified
  • Latest Bluetooth v4.0, GPS, Wi-Fi and wireless LAN 802.11 b/g/n tri-mode radios
  • 4 ft drop tested
  • Makes RFID tag reading simple and easy
  • Built on the Android mobile platform, the PA720 Android handheld device
  • Fully compatible with 3M™ Safety and Inspection Manager
  • 3M Catalog# PA720
TRUSTED_SUSTAINABILITY
Specifications
Product Type Handheld Computer
Model PA720