Learn More
Unity Lab Services™ Preferred Pipette Support Plan
GREENER_CHOICE
Catalog No. 12102114 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Multi-channel Pipette
Single-channel Pipette
Calibration Points:
2 x 10 Meaurements
2 x 5 Meaurements
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Calibration Points
12-102-114 Single-channel Pipette 2 x 10 Meaurements
12-102-112 Single-channel Pipette 2 x 5 Meaurements
12-102-113 Multi-channel Pipette 2 x 5 Meaurements
12-102-115 Multi-channel Pipette 2 x 10 Meaurements
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-102-114 Supplier Unity Lab Services™ Supplier No. PMC2X10DSC
Add to Cart
Add to Cart
Fast, quality-minded service that meets your diverse needs
Whether cGLP, cGMP, critical research, academia, or any special requirements you may need, our experienced service personnel are highly trained in all pipette brands and models. We provide calibration and a detailed 15-point pipette inspection of single and multichannel manual and electronic components to identify potential concerns that can impact pipettes from performing optimally. You will feel confident that your pipettes will have the accuracy and performance you require, when you need them.
Unity Pipette Services offers:
  • Service for any pipette brand and model
  • Reliable service options by our highly trained technicians
  • Fast and convenient prepaid pipette shipping packages
The Preffered Preventive Maintenance and Calibration Plan provides a thorough inspection, preventive maintenance and calibration. This plan includes a calibration certificate and sticker.
Ideal for customers who require:
  • Pipette inspection, liquid leak test, routine preventive maintenance and minor repairs
  • Do not need as-received data
  • Calibration
† Unity Lab Services™ provides a detailed 15-point pipette inspection of the following single and multi-channel manual and electronic components to identify potential concerns that can impact pipettes from performing optimally:
Checked routinely: Pipette body and display, tip ejection and ejector mechanism, tip cone(s), piston(s) assembly, o-ring(s), springs
Checked as applicable: Plunger bottom, plunger rod, volume lock, shaft coupling nut, seal(s), lubrication, battery recharging, electronics functionality, motor movement
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Single-channel Pipette
Calibration Points 2 x 10 Meaurements
Product Type Preferred Pipette Support Plan
Service Offering Preventive Maintenance and Calibration, 2 x 10