Learn More
Unity Lab Services™ Premium Pipette Support Plan
GREENER_CHOICE
Catalog No. 12102121
Change view
Click to view available options
For Use With (Equipment):
Multi-channel Pipette
Single or Multi-channel Pipette
Single-channel Pipette
Service Offering:
GxP Preventive Maintenance and Calibration, 3 x 10
GxP Preventive Maintenance and Calibration, 3 x 5
Repair Labor Charge Depot Service
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Service Offering
12-102-121 Single-channel Pipette GxP Preventive Maintenance and Calibration, 3 x 5
12-102-122 Multi-channel Pipette GxP Preventive Maintenance and Calibration, 3 x 5
12-102-124 Multi-channel Pipette GxP Preventive Maintenance and Calibration, 3 x 10
12-102-123 Single-channel Pipette GxP Preventive Maintenance and Calibration, 3 x 10
12-102-129 Single or Multi-channel Pipette Repair Labor Charge Depot Service
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 12-102-121 Supplier Unity Lab Services™ Supplier No. GXP3X5DSC
Add to Cart
Add to Cart
Fast, quality-minded service that meets your diverse needs
Whether cGLP, cGMP, critical research, academia, or any special requirements you may need, our experienced service personnel are highly trained in all pipette brands and models. We provide calibration and a detailed 15-point pipette inspection of single and multichannel manual and electronic components to identify potential concerns that can impact pipettes from performing optimally. You will feel confident that your pipettes will have the accuracy and performance you require, when you need them.
Unity Pipette Services offers:
  • Service for any pipette brand and model
  • Reliable service options by our highly trained technicians
  • Fast and convenient prepaid pipette shipping packages
Premium – GxP Preventive Maintenance and Calibration
The GxP Preventive Maintenance and Calibration Plan provides a comprehensive service that includes gravimetric evaluation of your pipette's performance prior to any inspection, preventive maintenance or minor repairs. This plan includes a calibration certificate and sticker.
Ideal for customers who require:
  • As-received data
  • Pipette inspection, liquid leak test, routine preventive maintenance and minor repairs
  • Calibration
  • Work in a highly regulated environment
  • Reduced OOTs (Out of Tolerance) that could invalidate research
† Unity Lab Services™ provides a detailed 15-point pipette inspection of the following single and multi-channel manual and electronic components to identify potential concerns that can impact pipettes from performing optimally:
Checked routinely: Pipette body and display, tip ejection and ejector mechanism, tip cone(s), piston(s) assembly, o-ring(s), springs
Checked as applicable: Plunger bottom, plunger rod, volume lock, shaft coupling nut, seal(s), lubrication, battery recharging, electronics functionality, motor movement
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Single-channel Pipette
Calibration Points 3 x 5 Measurements
Product Type Preventive Maintenance and Calibration
Service Offering GxP Preventive Maintenance and Calibration, 3 x 5
Includes Gravimetric evaluation of pipettes performance prior to any inspection, preventive maintenance or minor repairs; includes calibration certificate and sticker