Learn More
Caron Products And Services UNIV ANALOG OUTPUT 0-5V,4-20MA
Catalog No. NC1976385
Encompass_Preferred
Universal analog output (0-5V,4-20mA) for use with Caron 7400 series incubators. Provides three connections formonitoring temperature, humidity, and CO2. Factory installedat the time of order or can be retro-fitted in the field by aqualified technician.
Catalog No. NC1976385 Supplier Caron Products And Services Supplier No. OUTP303
Universal analog output (0-5V,4-20mA) for use with Caron 7400 series incubators. Provides three connections formonitoring temperature, humidity, and CO2. Factory installedat the time of order or can be retro-fitted in the field by aqualified technician.