Learn More
MSC Panduit Universal Circuit Breaker Lockout, Set Screw Style, Nylon, 1 ea
Catalog No. 5000684083
Encompass
The Universal Circuit Breaker Lockout Device is designed to isolate electrical energy sources for enhanced workplace safety and compliance with OSHA 1910.147. This set screw style device effectively locks out a wide range of single and multi-pole circuit breakers to prevent accidental or intentional energization during maintenance.
  • Constructed from durable nylon material
  • Accommodates breaker handles 0.30 in. to 0.60 in. high and 0.16 in. to 0.35 in. thick
  • Features a set screw design for a firm, secure fit against the handle
  • Accommodates one padlock to restrict access to the set screw
  • Designed for industrial and harsh application environments
  • Provides clear visual identification with red coloring
Catalog No. 50-006-84083 Supplier MSC Supplier No. 39387691
The Universal Circuit Breaker Lockout Device is designed to isolate electrical energy sources for enhanced workplace safety and compliance with OSHA 1910.147. This set screw style device effectively locks out a wide range of single and multi-pole circuit breakers to prevent accidental or intentional energization during maintenance.
  • Constructed from durable nylon material
  • Accommodates breaker handles 0.30 in. to 0.60 in. high and 0.16 in. to 0.35 in. thick
  • Features a set screw design for a firm, secure fit against the handle
  • Accommodates one padlock to restrict access to the set screw
  • Designed for industrial and harsh application environments
  • Provides clear visual identification with red coloring