Learn More
Chemglass Life Sciences 60mL Filter Funnel, Jacketed, Coarse Frit
SDP
Supplier:  Chemglass Life Sciences CG140306
Encompass_Preferred
60mL Filter Funnel, Jacketed, Coarse Frit. Similar to CG-1402 but with a water jacket enclosing the funnel bowl and upper portion of the stem. Jacket permits heating or cooling of the contents of the funnel during filtration. Stem length listed is the length below the jacket. Jacketed filter funnels are available in sizes other than those listed. Please call for details.
Catalog No. CG140306