Learn More
INDUSTRIAL MAGNETICS INC MAG-MATE J-HOOK WITH MX1500
SDP
Catalog No. NC1805437
Encompass
This item has a minimum order quantity of 3 cases per supplier requirements
MAG-MATE J-Hook With Mx1500 MX1500JH. Magnetic Universal J-Hooks. J-Hooks hold cables, wires, tubing, etc. to steel walls, ceilings or beams. Powerful magnet can be mounted on the hook in either of two positions for wall or ceiling installation.
Catalog No. NC1805437 Supplier INDUSTRIAL MAGNETICS INC Supplier No. MX1500JH
This item has a minimum order quantity of 3 cases per supplier requirements
MAG-MATE J-Hook With Mx1500 MX1500JH. Magnetic Universal J-Hooks. J-Hooks hold cables, wires, tubing, etc. to steel walls, ceilings or beams. Powerful magnet can be mounted on the hook in either of two positions for wall or ceiling installation.