Learn More
Premiere Magnesia UNIV SPILL SORBENT NEUTRALIZER
SDP
Catalog No. 501090655
Encompass_Preferred
Amphomag is a safe and user-friendly universal spill neutralizer and sorbent. It efficiently handles all of your spill control needs due to its unique ability to neutralize acids and bases. In addition, Amphomag contains an innovative multi-range pH indicator that further simplifies spill response. When applied to the spill, the color of Amphomag turns to red for acids and blue for bases. Keeping this one-size-fits-all product on hand eliminates the need to stock multiple reagents in your lab or facility. It also helps users avoid misapplication as it is universally compatible with most chemicals, creating a safer environment for you and your staff, and enabling quicker response time. Amphomag is available in 3 lb. shaker bottles, 50 lb. totes, and 50 lb. paper bags. 1309-48-4
Catalog No. 50-109-0655 Supplier Premiere Magnesia Supplier No. 398208
Amphomag is a safe and user-friendly universal spill neutralizer and sorbent. It efficiently handles all of your spill control needs due to its unique ability to neutralize acids and bases. In addition, Amphomag contains an innovative multi-range pH indicator that further simplifies spill response. When applied to the spill, the color of Amphomag turns to red for acids and blue for bases. Keeping this one-size-fits-all product on hand eliminates the need to stock multiple reagents in your lab or facility. It also helps users avoid misapplication as it is universally compatible with most chemicals, creating a safer environment for you and your staff, and enabling quicker response time. Amphomag is available in 3 lb. shaker bottles, 50 lb. totes, and 50 lb. paper bags. 1309-48-4