Learn More
SPEX CERTIPREP LLC 3P Universal Water Microbe Cocktail, 10 pellets
Catalog No. NC1728141
Encompass_Preferred
(NSI by Zeptometrix) A single pellet to QC all of your quantitative microbiology analyses! Formulated in our unique lyophilized flash dissolve pellet this microbiological standard contains multiple organisms that in aggregate allow the analyst to QC the following water microbiology tests at following approximate activity levels: Total Coliform 2400CFU/100mL, E.coli 1000CFU/100mL, Fecal Coliform 500CFU/100mL, P.aeruginosa 1000CFU/100mL, Enterococci 1000CFU/100mL, HPC 50CFU/mL.
Catalog No. NC1728141 Supplier SPEX CERTIPREP LLC 3P Supplier No. MICUNV10
(NSI by Zeptometrix) A single pellet to QC all of your quantitative microbiology analyses! Formulated in our unique lyophilized flash dissolve pellet this microbiological standard contains multiple organisms that in aggregate allow the analyst to QC the following water microbiology tests at following approximate activity levels: Total Coliform 2400CFU/100mL, E.coli 1000CFU/100mL, Fecal Coliform 500CFU/100mL, P.aeruginosa 1000CFU/100mL, Enterococci 1000CFU/100mL, HPC 50CFU/mL.