Learn More
Hach Company UniVer 3 Hardness Reagent, 454 g
Supplier:  Hach Company 21301H
Encompass
Combined indicator and buffer for determination of total hardness by titration methods. 454 g bottle.
Catalog No. 50-199-7396