Learn More
Hach Company UniVer 3 Hardness Reagent Powder Pillows, pk/100
Supplier:  Hach Company 96299
Encompass
Combined indicator/buffer for total hardness determination by titration. For use with total hardness test kits. Pack of 100 Powder Pillows.
Catalog No. 50-199-4477