Learn More
Hettich Centrifuges UNIVERSAL 320R, (12) KOVA,Package
SDP
Supplier:  Hettich Centrifuges 320RURINALYSIS
Encompass_Preferred
Hettich UNIVERSAL 320R URINALYSIS PACKAGE, (1) Centrifuge w/Temp. Control, (1) Horizontal Rotor, (4) Buckets, (4) Adapters KOVA, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7630
Add to Cart
Request a Quote