Learn More
Hettich Centrifuges UNIVERSAL 320R, (16) 5mL,Package
SDP
Catalog No. 502537632 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich UNIVERSAL 320R CONICAL TUBE PACKAGE 5mL BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets, (4) Adapters 5 mL Conical, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7632 Supplier Hettich Centrifuges Supplier No. 320RCONICAL5BC
Add to Cart
Add to Cart
Request a Quote
Hettich UNIVERSAL 320R CONICAL TUBE PACKAGE 5mL BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Rotor, (4) Buckets, (4) Adapters 5 mL Conical, (4) Bio-containment Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years