Learn More
Thomson Instrument Company Universal 4-Position Rack for Well Plates
SDP
Catalog No. 5000850163
Encompass_Preferred
The Thomson Universal 4-Position Rack for Well Plates holds up to 4 of Thomson s 6- 24- and 96-well plates for easy plate handling storage and transport Its compact durable design keeps plates secure during incubator shaking (up to 225 rpm) It is also compatible with standard 50 mL conical centrifuge tubes/mini-bioreactor tubes when placed in appropriate holder Whether moving plates between workstations or between a liquid handler and shaker the Universal 4-Position Rack is a practical accessory for labs using Thomson s well plates and 50 mL tubes
Catalog No. 50-008-50163 Supplier Thomson Instrument Company Supplier No. 1212605399
The Thomson Universal 4-Position Rack for Well Plates holds up to 4 of Thomson s 6- 24- and 96-well plates for easy plate handling storage and transport Its compact durable design keeps plates secure during incubator shaking (up to 225 rpm) It is also compatible with standard 50 mL conical centrifuge tubes/mini-bioreactor tubes when placed in appropriate holder Whether moving plates between workstations or between a liquid handler and shaker the Universal 4-Position Rack is a practical accessory for labs using Thomson s well plates and 50 mL tubes