Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ Socorex™ In-line™ Universal 337 Pipette Stand
SDP

Catalog No. 02912677 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Ice Blue
Mint Green
Pastel Rose
Vanilla Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02-912-677 Vanilla Yellow
02-912-675 Ice Blue
02-912-676 Mint Green
02-912-678 Pastel Rose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-912-677 Supplier DWK LIFE SCIENCES™ Supplier No. W851359
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Keep your workstation organized with the Universal 7 Pipette Work Station, Model 337. This universal stand holds up to 7 pipettes, fits all brands, and ensures proper storage. Available in multiple colors, with anti-slip pads for bench stability and an easy-to-clean design.

  • Space-saving pipette stand holds up to seven single-channel pipettes of most brands
  • Made of sturdy polyamid material, easy to clean
  • Heavy bottom plate and anti-slip pads guarantee bench stability
  • The work station fits new and old generations of Socorex and other brand pipettes
TRUSTED_SUSTAINABILITY

Specifications

Description Model 337
Autoclavable Not Autoclavable
Color Vanilla Yellow
Holds Pipets
Material Polyamide
Positions 7
Shape Round
Stackable No
Sterility Non-sterile
For Use With (Equipment) Univeral
No. of Racks 1 Each
Includes Anti-slip pads
Product Type Pipet Stand
Shelves None
Height (Metric) 220 mm
Width (Metric) 196 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.