Learn More
TRIBIOSCIENCE INC Universal AAV qPCR Titer Kit, 100 Tests
SDP
Catalog No. NC2887462
Encompass_Preferred
AAV qPCR FAST Titer Kit incorporates TribioScience FAST technology paradigm and Probe qPCR technology for an accurate, one-step, and time-saving qPCR titration of AAV serotypes. The probe is labeled with Fluorescence FAM.
Catalog No. NC2887462 Supplier TRIBIOSCIENCE INC Supplier No. TBS4054100
AAV qPCR FAST Titer Kit incorporates TribioScience FAST technology paradigm and Probe qPCR technology for an accurate, one-step, and time-saving qPCR titration of AAV serotypes. The probe is labeled with Fluorescence FAM.