Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Universal Absorbent Mat - Heavy Weight

Catalog No. 17111114 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
59 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-114 59 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-114 Supplier Brady™ Supplier No. SIR36
  • $616.00
    $434.65 / Each of 1

    Save $181.35

    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Constructed of durable, needle-punched polypropylene fibers, SIR Sorbent Industrial Rugs reduce slip hazards in high traffic areas, workstations and walkways.

  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
  • Heavy weight - specifically designed for continuous leaks and larger spills so you use less material
  • SIR Sorbent Industrial Rugs help eliminate slippery areas and unsightly tracking
  • Specially designed for use in high traffic areas

Specifications

Certifications/Compliance EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910.120(j)(1)(vii); OSHA 29 CFR 1910.22(a)(2)
Color Gray
Description Absorbent Mat
Capacity (English) 59 gal.
Capacity (Metric) 223.4 L
Material Polypropylene
Type Absorbent Mat
Packaging Quantity 300 ft./Roll
Dimensions (L x W) 150 ft. x 36 in.
Length (English) 300 ft.
Length (Metric) 91.44 m
Width (English) 36 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.