Learn More
Sigma Aldrich Fine Chemicals Biosciences Universal Antibody Dilutio
Catalog No. 502900449
Encompass
Ready to use buffer for dilution of antibodies in all immunoassays. This buffer does not contain any mammalian proteins phosphate sodium azide or mercury preservatives and can be used for dilution of all antibodies including peroxidase conjugates and antibodies to phosphoproteins. Not suitable for dilution of antibodies to S100 proteins.
Catalog No. 50-290-0449 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. U3635100ML
Ready to use buffer for dilution of antibodies in all immunoassays. This buffer does not contain any mammalian proteins phosphate sodium azide or mercury preservatives and can be used for dilution of all antibodies including peroxidase conjugates and antibodies to phosphoproteins. Not suitable for dilution of antibodies to S100 proteins.