Learn More
MSC 80/20 Inc. Standard Structural Pivot, Aluminum, 10 Series, 1 ea
Catalog No. 5000516918
Encompass
This universal pivot arm is designed for integration with 10 series framing systems and bolt kit 3393 to provide versatile movement for structural components. Manufactured from 6063-T6 aluminum with an anodized finish, this component offers reliable performance in industrial material handling applications.
  • Provides a 180° maximum range of motion
  • Constructed from durable 6063-T6 aluminum with an anodized finish
  • Includes necessary fasteners for assembly
  • ESD safe design
  • Compatible with 10 series profiles
Catalog No. 50-005-16918 Supplier MSC Supplier No. 06036370
This universal pivot arm is designed for integration with 10 series framing systems and bolt kit 3393 to provide versatile movement for structural components. Manufactured from 6063-T6 aluminum with an anodized finish, this component offers reliable performance in industrial material handling applications.
  • Provides a 180° maximum range of motion
  • Constructed from durable 6063-T6 aluminum with an anodized finish
  • Includes necessary fasteners for assembly
  • ESD safe design
  • Compatible with 10 series profiles