Learn More
NEOGEN Universal Beer Agar, 10 kg
SDP
Catalog No. 502015647
Encompass_Preferred
Universal Beer Agar is used for the cultivation of bacteria and yeasts encountered in the brewing industry in a laboratory setting. Universal Beer Agar is not intended for use in the diagnosis of disease or other conditions in humans. Universal Beer Agar was developed by Kozulis and Page and used as a basal medium supplemented with beer. The developers compared this formula with other media commonly used in breweries for detecting microbial contamination. The characteristics of Universal Beer Agar are closer to natural environmental conditions found in a typical brewery. Universal Beer Agar supports the growth of more lactic acid bacteria and yields larger colonies in a shorter time than traditional brewer’s media. Supplementing this medium with beer has produced a selective environment for organisms that are adapted to existent conditions in the brewery.For more information visit https://www.neogen.com/solutions/Microbiology/universal-beer-agar
Catalog No. 50-201-5647 Supplier NEOGEN Supplier No. 700003474
Add to Cart
Add to Cart
Request a Quote
Universal Beer Agar is used for the cultivation of bacteria and yeasts encountered in the brewing industry in a laboratory setting. Universal Beer Agar is not intended for use in the diagnosis of disease or other conditions in humans. Universal Beer Agar was developed by Kozulis and Page and used as a basal medium supplemented with beer. The developers compared this formula with other media commonly used in breweries for detecting microbial contamination. The characteristics of Universal Beer Agar are closer to natural environmental conditions found in a typical brewery. Universal Beer Agar supports the growth of more lactic acid bacteria and yields larger colonies in a shorter time than traditional brewer’s media. Supplementing this medium with beer has produced a selective environment for organisms that are adapted to existent conditions in the brewery.For more information visit https://www.neogen.com/solutions/Microbiology/universal-beer-agar