Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brew Plate™ UNIVERSAL BEER AGAR - 500G
SCI_ED

Catalog No. S72013
Change view
Click to view available options
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S72013 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72013 Supplier Brew Plate™ Supplier No. 61401404

Universal Beer Agar is used for culturing microorganisms of significance in the brewing industry.

Uses:

  • Culturing microorganisms

Specifications

Content And Storage 2-30° C
Product Type Agar
For Use With (Application) Cultivating yeast and bacteria found in brewing.
Quantity 1 EA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.