Learn More
Med Vet International Universal Bottle Adapter, Fits Bottle Openings 18-22mm, J0578, 5 Pack
Catalog No. 5000495305
Encompass_Preferred
This adaptor features a graduated rubber plug that fits bottle openings 18-22mm. The tethered plug lets you reseal the bottle for later use. Attach oral dose syringe and withdraw desired dose.Will not work as well with a luer tip syringe
Catalog No. 50-004-95305 Supplier Med Vet International Supplier No. JOR578
This adaptor features a graduated rubber plug that fits bottle openings 18-22mm. The tethered plug lets you reseal the bottle for later use. Attach oral dose syringe and withdraw desired dose.Will not work as well with a luer tip syringe