Learn More
Stoelting™ Universal Camera Mounting Bracket
SDP
Catalog No. 10000336 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-000-336 Neuroscience
1 options
Catalog No. 10-000-336 Supplier Stoelting™ Supplier No. 60545
 Restricted Item
Add to Cart
Add to Cart
Stoelting™ Universal Camera Mounting Bracket allows the versatility of wall, ceiling, or drop ceiling rail mounting.
Can be used as 3 in. mount with head attached to base or 6 in. mount when shaft is added.
Specifications
For Use With (Application) Neuroscience