Learn More
Sigma Aldrich Fine Chemicals Biosciences Universal carousel pipette
Catalog No. 502900025
Encompass
Pipettes never looked so good on your bench topHold and store pipettes in style with the new universal carousel pipette stand that holds up to 8 single channel 4 multi channel or a combination of both pipettes. A sleek and modern design enables this stand to securely hold virtually every known brand of pipette in the marketplace making it truly universal while adding a pop of color to a bench top. Inserts on the upper holders can be added or removed to accommodate the fit of different of pipette widths. One handed removal or replacement of pipettes is made easy with a rotating head and stable weighted base. Moving the stand while keeping pipettes in place is simple with a quick grip handle allowing a user to grab and go. The pipette stands come with additional upper leaves to enable multiple color coding configuration options as well as a change in color to your work space for a bit more fun.Ideal for labs that do not wish to be tied into a specific brand of pipette or alread
Catalog No. 50-290-0025 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS120499
Pipettes never looked so good on your bench topHold and store pipettes in style with the new universal carousel pipette stand that holds up to 8 single channel 4 multi channel or a combination of both pipettes. A sleek and modern design enables this stand to securely hold virtually every known brand of pipette in the marketplace making it truly universal while adding a pop of color to a bench top. Inserts on the upper holders can be added or removed to accommodate the fit of different of pipette widths. One handed removal or replacement of pipettes is made easy with a rotating head and stable weighted base. Moving the stand while keeping pipettes in place is simple with a quick grip handle allowing a user to grab and go. The pipette stands come with additional upper leaves to enable multiple color coding configuration options as well as a change in color to your work space for a bit more fun.Ideal for labs that do not wish to be tied into a specific brand of pipette or alread