Learn More
Cole-Parmer Cole-Parmer Ion Exchange Filter Cartridge, universal
Supplier:  Cole-Parmer 0150625
Encompass_Preferred
Design your system with any combination of four different cartridges that produces water equivalent to single-distilled water. The cartridges can purify water up to 15 M-cm.
Catalog No. NC9651811