Learn More
Carbosynth LLC. Ready-to-use biotin free reagent applicable for blocking ELISA plates
SDP
Supplier:  Carbosynth LLC. 85R108_R0.25L
Encompass_Preferred
Ready-to-use biotin free reagent applicable for blocking ELISA plates/membranes, diluting complex specimens and diverse detection reagents/conjugates including high performance and ultrasensitive biotin-streptavidin systems. Effectively eliminates NSB in ELISA, blotting, ICT and IHC applications.
Catalog No. 50-125-3502