Learn More
Biochain Institute Inc Universal cDNA Reverse Transcribed by Random Hexamer: Dog Normal Tissues, 100 rxn/PK
SDP
Catalog No. 50184212
Encompass_Preferred
BioChain's Dog Universal cDNA is prepared from major organs of both male and female Beagle dogs. This cDNA serves as a standard for comparison of gene expression by RT-PCR or as a gene pool for cloning dog genes.
Catalog No. 50-184-212 Supplier Biochain Institute Inc Supplier No. C4734565R
BioChain's Dog Universal cDNA is prepared from major organs of both male and female Beagle dogs. This cDNA serves as a standard for comparison of gene expression by RT-PCR or as a gene pool for cloning dog genes.