Learn More
Biochain Institute Inc Universal cDNA Reverse Transcribed by Oligo dT Primer: Human Normal Tissues, 100 rxn/PK
SDP
Catalog No. 50184213
Encompass_Preferred
BioChain's Human Universal cDNA is prepared from major organs of both male and female adult donors. This cDNA serves as a standard for comparison of gene expression by RT-PCR or as a gene pool for cloning human genes.
Catalog No. 50-184-213 Supplier Biochain Institute Inc Supplier No. C4234565
BioChain's Human Universal cDNA is prepared from major organs of both male and female adult donors. This cDNA serves as a standard for comparison of gene expression by RT-PCR or as a gene pool for cloning human genes.