Learn More
Biochain Institute Inc Universal cDNA Reverse Transcribed by Random Hexamer: Human Normal Tissues, 100 rxn/PK
SDP
Catalog No. 50184214
Encompass_Preferred
BioChain's Human Universal cDNA is prepared from major organs of both male and female normal adult donors. This cDNA is ready to use for comparison of gene expression by RT-PCR or as a gene pool for cloning human genes. It is a pool from 2 male donors (64 and 71 years old) and 3 female donors (32, 69, and 76 years old).
Catalog No. 50-184-214 Supplier Biochain Institute Inc Supplier No. C4234565R
BioChain's Human Universal cDNA is prepared from major organs of both male and female normal adult donors. This cDNA is ready to use for comparison of gene expression by RT-PCR or as a gene pool for cloning human genes. It is a pool from 2 male donors (64 and 71 years old) and 3 female donors (32, 69, and 76 years old).