Learn More
Biochain Institute Inc Universal cDNA Reverse Transcribed by Oligo dT Primer: Monkey Normal Tissues, 100 rxn/PK
SDP
Catalog No. 50184215
Encompass_Preferred
BioChain's Monkey Universal cDNA is prepared from major organs of both male and female Cynomolgus and Rhesus monkeys. This cDNA serves as a standard for comparison of gene expression by RT-PCR or as a gene pool for cloning monkey genes.
Catalog No. 50-184-215 Supplier Biochain Institute Inc Supplier No. C4534565
BioChain's Monkey Universal cDNA is prepared from major organs of both male and female Cynomolgus and Rhesus monkeys. This cDNA serves as a standard for comparison of gene expression by RT-PCR or as a gene pool for cloning monkey genes.