Learn More
Biochain Institute Inc Universal cDNA Reverse Transcribed by Oligo dT Primer: Mouse Normal Tissues, 100 rxn/PK
SDP
Catalog No. 50184217
Encompass_Preferred
BioChains' Mouse Universal cDNA is prepared from several male and female Balb/C mice whole bodies without fur. This cDNA serves as a standard for comparison of gene expression by RT-PCR or as a gene pool for cloning mouse genes.
Catalog No. 50-184-217 Supplier Biochain Institute Inc Supplier No. C4334566
BioChains' Mouse Universal cDNA is prepared from several male and female Balb/C mice whole bodies without fur. This cDNA serves as a standard for comparison of gene expression by RT-PCR or as a gene pool for cloning mouse genes.