Learn More
Biochain Institute Inc Universal cDNA Reverse Transcribed by Oligo dT Primer: Rat Normal Tissues, 100 rxn/PK
SDP
Catalog No. 50184219
Encompass_Preferred
BioChains' Rat Universal cDNA is prepared from several male and female Sprague Dawley or Wistar rat whole bodies without fur. This cDNA serves as a standard for comparison of gene expression by RT-PCR or as a gene pool for cloning rat genes.
Catalog No. 50-184-219 Supplier Biochain Institute Inc Supplier No. C4434567
BioChains' Rat Universal cDNA is prepared from several male and female Sprague Dawley or Wistar rat whole bodies without fur. This cDNA serves as a standard for comparison of gene expression by RT-PCR or as a gene pool for cloning rat genes.