Learn More
Arrow Engineering PHC-625 Power Hold Clamp
Catalog No. NC1721856
Encompass_Preferred
The Arrow Mixing Products PHC-625 fits support stands from 3/8″ to 5/8″ dia. (9.5 mm to 16 mm). Designed to securely clamp laboratory mixers to support stands the PHC-625 is strong, reliable, and built to provide a firm grip even under demanding conditions. Ideal for both light and heavy-duty mixing tasks, it is engineered for versatility and fits multiple types of support clamp stands with ease.
Catalog No. NC1721856 Supplier Arrow Engineering Supplier No. PHC625
The Arrow Mixing Products PHC-625 fits support stands from 3/8″ to 5/8″ dia. (9.5 mm to 16 mm). Designed to securely clamp laboratory mixers to support stands the PHC-625 is strong, reliable, and built to provide a firm grip even under demanding conditions. Ideal for both light and heavy-duty mixing tasks, it is engineered for versatility and fits multiple types of support clamp stands with ease.