Learn More
Chemglass Life Sciences Universal Clip Plates, 2/Pk (Fig. 10)
SDP
Catalog No. 5000083203
Encompass
Combines rocking and rotating in one compact and versatile unit. Easily converts to rocking to rotating (or vice versa) in just 5 seconds. Magnetic stainless steel platform and clip plate System holds almost any container at any angle and expansion kits available for increased capacity and different containers. Magnetic Strips hold bags for complete and even soaking of membrane-based blot assays/washings. Ideal for Cold Rooms and Incubators and Maintains set speed between 0-38 degreesC. Optional 3-D orbital shaker attachment. Supplied with 1 clip plate each for 10-13 mm, 15-17mm, 28-30mm tubes, 4 bag mounting strips and a non-skid tray.
Catalog No. 50-000-83203 Supplier Chemglass Life Sciences Supplier No. CLS3960013
Combines rocking and rotating in one compact and versatile unit. Easily converts to rocking to rotating (or vice versa) in just 5 seconds. Magnetic stainless steel platform and clip plate System holds almost any container at any angle and expansion kits available for increased capacity and different containers. Magnetic Strips hold bags for complete and even soaking of membrane-based blot assays/washings. Ideal for Cold Rooms and Incubators and Maintains set speed between 0-38 degreesC. Optional 3-D orbital shaker attachment. Supplied with 1 clip plate each for 10-13 mm, 15-17mm, 28-30mm tubes, 4 bag mounting strips and a non-skid tray.