Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Universal Cone Adapter

Catalog No. 19169585 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-169-585 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-169-585 Supplier Accuform™ Supplier No. FBC107OR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accuform™ Universal Cone Adapter provides a simple, convenient solution to attach signs and other safety products to standard traffic cone tops

  • Durable plastic construction
  • Fits most standard traffic cone top
  • Unique design makes it easy to mount signs, flags, and lights
  • Adapter can also be used with rope or barricade tape

Specifications

For Use With (Equipment) Standard Traffic Cones
For Use With (Application) Sign Mounting
Material Plastic
Color Orange
Product Type Cone Top Adapter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.