Learn More
Perkin Elmer US LLC Universal Connector, Pkg. 5
Supplier:  Perkin Elmer US LLC N9302149
Encompass_Preferred
The new fused-silica tapered connector design is easy to use and allows connection of 0.1 mm I.D. to 0.53 mm I.D. columns.
Catalog No. 50-905-2325