Learn More
Perkin Elmer US LLC Universal Data Acquisition Standards Kit
Catalog No. 502104640
Encompass_Preferred
Kit Includes one 125mL bottle of each: Part no N9300232 Part no. N9300233 Part no. N9300234 Part no. N9300253* Part no. N9300235 10 ug/mL 10 ug/mL 10 ug/mL 10 ug/mL 10 ug/mL Ce Al Au Hg B Dy Ag Hf Ge Er As Ir Mo Eu Ba Pd Nb Gd Be Pt P Ho Bi Rh Re La Ca Ru S Lu Cd Te Si Nd Co Sb Ta Pr Cr Sn Ti Sm Cs W Sc Cu Zr Tb Fe Th Ga Tm In Y K Yb Li Mg Mn Na Ni Pb Rb Se Sr Ti V U Zn
Catalog No. 50-210-4640 Supplier Perkin Elmer US LLC Supplier No. N9306225
Kit Includes one 125mL bottle of each: Part no N9300232 Part no. N9300233 Part no. N9300234 Part no. N9300253* Part no. N9300235 10 ug/mL 10 ug/mL 10 ug/mL 10 ug/mL 10 ug/mL Ce Al Au Hg B Dy Ag Hf Ge Er As Ir Mo Eu Ba Pd Nb Gd Be Pt P Ho Bi Rh Re La Ca Ru S Lu Cd Te Si Nd Co Sb Ta Pr Cr Sn Ti Sm Cs W Sc Cu Zr Tb Fe Th Ga Tm In Y K Yb Li Mg Mn Na Ni Pb Rb Se Sr Ti V U Zn