Learn More
Perkin Elmer US LLC Universal Demountable Cell Replacement Plug - Qty. 6
Catalog No. 502113502
Encompass_Preferred
The Universal Demountable Cell comes standard in various IR Sampling kits. This cell is an assembled cell, allowing you to customize the window material and path length to meet your current needs.
Catalog No. 50-211-3502 Supplier Perkin Elmer US LLC Supplier No. 04821008
The Universal Demountable Cell comes standard in various IR Sampling kits. This cell is an assembled cell, allowing you to customize the window material and path length to meet your current needs.