Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Universal DNA Extraction Buffer
SDPSCI_ED

Catalog No. s94632 Shop All EDVOTEK Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S94632 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94632 Supplier Edvotek™ Supplier No. 627
Only null left
Add to Cart
Add to Cart

Recommended for the extraction of DNA from various fruit and vegetable tissues.

  • Composition is safe for classroom use
  • Ideal for use in developing independent inquiry-based experiments
  • Class size: 50 extractions

Specifications

Format Bottle
For Use With (Application) Extraction of DNA from various fruit and vegetable tissues and developing independent inquiry-based experiments
No. of Reactions 50
Product Type DNA Extraction Buffer
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.