Learn More
AVANTOR MASTERFLEX (VWR LLC) Universal Drum Wrench, Dual-Ended, Non-Sparking Brass
Catalog No. NC1362332
Encompass
Use the round end for outside splines of bung caps. Use the long end for internal splines of bung caps.
Catalog No. NC1362332 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7060791
Use the round end for outside splines of bung caps. Use the long end for internal splines of bung caps.