Learn More
Electron Microscopy Sciences R-Universal Epitope Recovery Buffer (10x stock) 500 ML
SDP
Catalog No. 501926580
Encompass_Preferred
EMS UNIVERSAL Epitope Recovery Buffer is a novel (patent-pending) technology for epitope recovery, primarily based on reversing the fixation effects of formaldehyde, which created links primarily between ε-amino groups of lysine and other amino-groups. In extensive testing the buffer has shown excellent results when used with different antibodies, including those that require low or high pH buffers or protolithic treatment for successful staining.
The EMS Universal Buffer may be used in any heat-induced epitope recovery system (the time and temperature of treatment should be tested), but was specifically adjusted for tissue sections processing in our Antigen Retriever (62700 Series). Using our Universal Buffer in our Retriever guarantees the highest rate of success in recovering epitope for any antibody, especially one that was never previously used on formalin-fixed sections. Shelf Life: 12 months.
Catalog No. 50-192-6580 Supplier Electron Microscopy Sciences Supplier No. 6271920
EMS UNIVERSAL Epitope Recovery Buffer is a novel (patent-pending) technology for epitope recovery, primarily based on reversing the fixation effects of formaldehyde, which created links primarily between ε-amino groups of lysine and other amino-groups. In extensive testing the buffer has shown excellent results when used with different antibodies, including those that require low or high pH buffers or protolithic treatment for successful staining.
The EMS Universal Buffer may be used in any heat-induced epitope recovery system (the time and temperature of treatment should be tested), but was specifically adjusted for tissue sections processing in our Antigen Retriever (62700 Series). Using our Universal Buffer in our Retriever guarantees the highest rate of success in recovering epitope for any antibody, especially one that was never previously used on formalin-fixed sections. Shelf Life: 12 months.